Learn More
Brady™ Aggressive Adhesive Die-cut Multipurpose Nylon Label Tape for M6/M7 Printers
Aggressive adhesive and high pliability are the hallmarks of this nylon cloth label material designed to meet a broad array of identification requirements, from control panels to liquid nitrogen environments.
38.70 EUR - 127.00 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Halogen-free (DIN VDE 0472 Part 815) |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | White |
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | |||
17858124
|
Brady™
173311 |
0.5 x 0.5 in. | M710, BMP71 | 12.7 mm | Square |
73.15 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17838454
|
Brady™
174349 |
0.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Square |
79.05 EUR
Pack of 500 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||
17818844
|
Brady™
173395 |
1 x 1 in. | M710, BMP71 | 25.4 mm | Square |
59.70 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17828404
|
Brady™
174225 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Square |
64.50 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17868974
|
Brady™
174594 |
0.25 x 1.5 in. | M710, BMP71 | 6.35 mm | Rectangle |
77.65 EUR
Pack of 750 |
In Stock
Log in to see stock. |
|||
17878194
|
Brady™
173307 |
0.25 x 1.9 in. | M710, BMP71 | 6.35 mm | Rectangle |
89.60 EUR
Pack of 750 |
In Stock
Log in to see stock. |
|||
17811389
|
Brady™
173790 |
0.25 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
75.75 EUR
Pack of 750 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||
17888284
|
Brady™
174357 |
0.25 x 1.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 6.35 mm | Rectangle |
95.85 EUR
Pack of 750 |
In Stock
Log in to see stock. |
|||
17838864
|
Brady™
174650 |
0.5 x 0.9 in. | M710, BMP71 | 12.7 mm | Rectangle |
117.00 EUR
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
17818384
|
Brady™
174370 |
0.5 x 0.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
127.00 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17888294
|
Brady™
174184 |
0.75 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
72.15 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17818464
|
Brady™
174218 |
0.75 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
59.60 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17801499
|
Brady™
174273 |
0.75 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
76.10 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17868194
|
Brady™
174201 |
1 x 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
86.20 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17879024
|
Brady™
173415 |
1 x 0.375 in. | M710, BMP71 | 25.4 mm | Rectangle |
79.80 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17878784
|
Brady™
173401 |
1 x 0.75 in. | M710, BMP71 | 25.4 mm | Rectangle |
55.20 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17898364
|
Brady™
174103 |
1 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
52.10 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17888114
|
Brady™
174280 |
1 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
82.10 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17898254
|
Brady™
174354 |
1 x 1.9 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
123.00 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17888434
|
Brady™
173310 |
1 x 1.9 in. | M710, BMP71 | 25.4 mm | Rectangle |
115.00 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17818924
|
Brady™
173355 |
1.5 x 0.5 in. | M710, BMP71 | 38.1 mm | Rectangle |
74.35 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17828844
|
Brady™
174265 |
1.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm | Rectangle |
80.35 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17888904
|
Brady™
173347 |
1.5 x 0.75 in. | M710, BMP71 | 38.1 mm | Rectangle |
78.75 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17898924
|
Brady™
173340 |
1.5 x 1 in. | M710, BMP71 | 38.1 mm | Rectangle |
76.00 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17831379
|
Brady™
173769 |
1 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
53.10 EUR
Pack of 100 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||
17868344
|
Brady™
174086 |
1.25 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 31.75 mm | Rectangle |
97.60 EUR
Pack of 250 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||
17899034
|
Brady™
174575 |
2.5 x 1 in. | M710, BMP71 | 63.5 mm | Rectangle |
44.45 EUR
Pack of 100 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||
17891569
|
Brady™
173756 |
2.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 63.5 mm | Rectangle |
43.40 EUR
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||
17888464
|
Brady™
174085 |
0.375 x 0.825 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
38.70 EUR
Pack of 500 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||
17888734
|
Brady™
173427 |
0.75 x 0.5 in. | M710, BMP71 | 19.05 mm | Rectangle |
66.80 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
Description
- Highly pliable material conforms tightly to curved surfaces such as wires and cables while also adhering to stainless steel, textured ABS, polypropylene and many other material types
- Resists exposure to cold temperatures, chemicals, greases, oils, cleaners, and detergents
- Low profile label material adheres to smooth, rough, or powder coated surfaces or even materials with low surface energy
- UL Recognized to UL969, CSA to C222 No 015-95, RoHS compliant to 2011/65/EU (Pass 2015/863)
- Made of B-499 nylon cloth material
Specifications
Autoclavable | |
Permanent Acrylic | |
White | |
Die-cut | |
Single-side Printable | |
Labels | |
Component and Equipment Labeling, General Identification, Laboratory Labeling | |
-40°F to +194°F | |
0.0065 in. |
Halogen-free (DIN VDE 0472 Part 815) | |
None | |
Matte | |
Nylon | |
Thermal Transfer | |
Non-sterile | |
No | |
-40°C to +90°C | |
0.166 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ Aggressive Adhesive Die-cut Multipurpose Nylon Label Tape for M6/M7 Printers