Learn More
Al-23 Tube, Both Ends Open, O.D.: 40mm, I.D.: 34mm, Thermo Scientific™
69.72 EUR - 568.00 EUR
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374017
View Documents
Thermo Scientific Alfa Aesar
031577.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11384017
View Documents
Thermo Scientific Alfa Aesar
031577.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11334027
View Documents
Thermo Scientific Alfa Aesar
031577.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11394017
View Documents
Thermo Scientific Alfa Aesar
031577.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314027
View Documents
Thermo Scientific Alfa Aesar
031577.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11324027
View Documents
Thermo Scientific Alfa Aesar
031577.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Al-23 Tube, Both Ends Open, O.D.: 40mm, I.D.: 34mm, Thermo Scientific™