Learn More
Thermo Scientific™ Ala-Leu-Ala-Leu
For cellular and molecular biology applications
Brand: Thermo Scientific™ J66857.M^
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
HPLC | |
-30°C to -10°C | |
386.49 | |
1 mg | |
A-L-A-L |
ALAL Linker Peptide | |
Solid | |
Ala-Leu-Ala-Leu | |
RUO - research use only |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only