Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
DESIGN FEATURES:
- Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
- Unexpected leakages spills or other releases
- Loading and unloading trucks and vehicles
- Loading trucks and vehicles
- Product inspection
- Cleaning of plant and machines
Specifications
Specifications
Type | Chemical-resistant Apron |
Size | 4X-Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Disposable | Reusable |
Closure Type | Hook and Loop |
Sterility | Non-sterile |
Weight (Metric) | 80 gsm |
Seam Type | Welded |
Certifications/Compliance (Europe) | CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126) |
Product Content Correction
Product Title
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01 >
Spot an opportunity for improvement?Share a Content Correction