Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Aluminum RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1784140
Description
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Canister | |
RD40 | |
Aluminum alloy with PE cover activated and impregnated charcoal. | |
Respiratory protection against mercury vapors, solid and liquid particles | |
Model HgP3 |
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 757, EC Attestation- EC Attestation, EC Attestation Number-757 | |
Red, White | |
ALU RD40 HG+P3 | |
Full-Face mask equipped with an Rd40 connector. | |
1 Ea. |