Description
- Highest available attenuation rated earplug at 39dB
- Smooth tapered finish makes fitting easier and helps form a comfortable fit
- Flange shape, when rolled down, serves as a very effective insertion device
- Ideal for those looking for higher attenuation
Specifications
Specifications
Color | Neon Green |
Quantity | 200/Pk. |
Corded | Uncorded |
Product Type | Ear Plugs |
Style | Roll-down |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ E-A-Rsoft FX Earplugs >
Spot an opportunity for improvement?Share a Content Correction