Learn More
Eppendorf™ Fixed Angle High Speed Rotor
For use in 5430 Micro Centrifuges.
Brand: Eppendorf™ 5427710000
732.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes aerosol-tight lid
- PTFE coated
- Holds 24 x 1.5/2.0mL 30.000 x g safe micro test tubes
Specifications
Aluminum | |
24 x 1.5/2 mL | |
17,500 rpm |
High Speed Rotor | |
30,130 x g | |
5430 Microcentrifuge |
Product Content Correction
Product Title
Eppendorf™ Fixed Angle High Speed Rotor > Max Speed: 17500rpm
Spot an opportunity for improvement?Share a Content Correction