Learn More
Wheaton™ Aluminum/Polypropylene Flip Tear Seal
Brand: Wheaton™ 700103B
193.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Blue | |
Aluminum, Polypropylene | |
Closure | |
20 mm |