missing translation for 'onlineSavingsMsg'
Learn More
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11359927

Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11359927
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11359927

Brand: Thermo Scientific Alfa Aesar 042259.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 250 g
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Weight ≈0.14g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA08U033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042259.30,042259.18
Lot NumberB05M08 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042259.30,042259.18
Lot NumberH18X038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number042259.30,042259.18
Lot NumberL23T056 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042259.30,042259.18
Lot NumberA24Y016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042259.30,042259.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.