Learn More
WTW™ Ammonia and Ammonium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 250426
235.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description

Specifications
Glyoxal-bis-hydroxyanil | |
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers | |
100 | |
100 Tests | |
Calcium (Ca) reagent test kit |
For waste water, drinking water, sea water | |
Certificates | |
0.010 to 3 mg/L NH4-N, 0.000 to 0.730 mg/L NH3; (pH 8.5 at 25°C) | |
Photometric |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- ammonium test method
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, 1-hydroxyethane-1,1-diphosphonic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.