missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Dichromate, 99.5+%, Extra Pure, SLR, Fisher Chemical™

108.00 EUR - 230.00 EUR

Chemical Identifiers

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
MDL Number 10879
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12675157
View Documents
Fisher Chemical
A/4420/50
250 g N/A N/A N/A
12685157
View Documents
Fisher Chemical
A/4420/53
500 g N/A N/A N/A
12695157
View Documents
Fisher Chemical
A/4420/60
1 kg N/A N/A N/A
Description

Description

Specifications

Chemical Identifiers

7789-09-5
252.06
JOSWYUNQBRPBDN-UHFFFAOYSA-P
Cr2H8N2O7
10879

Specifications

170°C
Orange
Solid
10879
252.06
Plastic powder jar
7789-09-5
3 to 4
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
252.07g/mol
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Dichromate, 99.5+%, Extra Pure, SLR, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.