missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydrogen Difluoride, 95%, Reagent grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | 125°C |
Color | White |
Quantity | 2.5 kg |
Linear Formula | NH4HF2 |
UN Number | UN1727 |
Formula Weight | 57.04g/mol |
Percent Purity | 95% |
Grade | Reagent |
Packaging | Poly Bottle |
Appearance | Crystals or flakes |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Hydrogen Difluoride, 95%, Reagent grade, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction