missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

32.86 EUR - 85.22 EUR

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium bifluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324006
SDS View Documents
Thermo Scientific Alfa Aesar
L17237.22
100 g
32.86 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
11334006
SDS View Documents
Thermo Scientific Alfa Aesar
L17237.36
500 g
37.08 EUR
500g
Please to purchase this item. Need a web account? Register with us today!
11344006
SDS View Documents
Thermo Scientific Alfa Aesar
L17237.0E
2500 g
85.22 EUR
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Especificaciones
Show More
Videos
Seguridad y manipulación
Show More
SDS
Documentos

Documentos

Certificados

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado