missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMPK A2/B1/G1, GST Tag, His Tag Recombinant Protein
Human AMPK A2/B1/G1, GST Tag, His Tag Recombinant Protein
1052.00 EUR
Specifications
Accession Number | NP_0062243.2 I NP_006244.2 I NP_002724.1 |
---|---|
For Use With (Application) | Kinase Assay |
Gene Alias | AMPK A2/B1/G1 |
KinaseFamily | CAMKL Kinase Family |
Species | Human |
Description
AMP-activated protein kinase (AMPK) is a heterotrimeric complex that acts as a sensor of cellular energy levels. The signaling cascades initiated by activating AMPK are critical to regulating metabolic events in the liver, skeletal muscle, heart, adipose tissue, and pancreas. An impairment in fuel metabolism that occurs in obesity is a factor leading to Type 2 diabetes. The insulin resistance associated with type 2 diabetes is most profound at the level of skeletal muscle, the primary site of glucose and fatty acid utilization. Activation of AMPK is of interest for the treatment of type 2 diabetes.Specifications
NP_0062243.2 I NP_006244.2 I NP_002724.1 | |
AMPK A2/B1/G1 | |
Human | |
GST Tag, His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus |
Kinase Assay | |
CAMKL Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.02% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |