missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anillin Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68216UV
This item is not returnable.
View return policy
Description
Anillin Polyclonal antibody specifically detects Anillin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Anillin | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
actin-binding protein anillin, ANILLIN, anillin (Drosophila Scraps homolog), actin binding protein, anillin, actin binding protein, anillin, actin binding protein (scraps homolog, Drosophila), DKFZp779A055, Drosophila Scraps homolog, scra, Scraps | |
The immunogen recognized by this antibody maps to a region between residue 1074 and 1124 of human actin binding protein Anillin using the numbering given in entry NP_061155.2 (GeneID 54443). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54443 | |
Store at 4C in the dark. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Anillin Antibody, DyLight 350, Novus Biologicals™ > 0.1 mL; DyLight 350
Spot an opportunity for improvement?Share a Content Correction