Learn More
phosphorylase kinase, gamma 1 (muscle), Mouse, Clone: 2B4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PHKG1.
Brand: Abnova H00005260-M02.100ug
Description
This gene is a member of the Ser/Thr protein kinase family and encodes a protein with one protein kinase domain and two calmodulin-binding domains. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. This gene has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12. [provided by RefSeq
Sequence: MTRDEALPDSHSAQDFYENYEPKEILGRGVSSVVRRCIHKPTSQEYAVKVIDVTGGGSFSPEEVRELREATLKEVDILRKVSGHPNIIQLKDTYETNTFFFLVFDLMKRGELFDYLTEKVTLSEKETRKIMRALLEVICTLHKLNIVHRDLKPENILLDDNMNIKLTDFGFSCQLEPGERLREVCGTPSYLAPEIIECSMNEDHPGYGKEVDMWSTGVIMYTLLAGSPPFWHRKQMLMLRMIMSGNYQFGSPEWDDYSDTVKDLVSRFLVVQPQNRYTAEEALAHPFFQQYLVEEVRHFSPRGKFKVIALTVLASVRIYYQYRRVKPVTREIVIRDPYALRPLRRLIDAYAFRIYGHWVKKGQQQNRAALFENTPKAVLLSLAEEDYSpecifications
phosphorylase kinase, gamma 1 (muscle) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC051327 | |
PHKG1 | |
PHKG1 (AAH51327, 1 a.a. ∼ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
2B4 | |
Mouse monoclonal antibody raised against a full length recombinant PHKG1. | |
PHKG1 | |
PHKG | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
5260 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.