Learn More
anti-Siglec-9, PE, Clone: 191240, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1139P-025

Description
Siglec-9 Monoclonal specifically detects Siglec-9 in Human samples. It is validated for Flow Cytometry.Specifications
Siglec-9 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27180 | |
Human | |
Purified |
Flow Cytometry | |
191240 | |
Flow Cytometry 10 uL/10^6 cells | |
Q9Y336 | |
SIGLEC9 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 | |
25 Tests | |
Primary | |
Detects human Siglec-9 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -7 or -10 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.