Learn More
Sartorius Anti-Static Devices for Balances
Control charges on samples
1890.00 EUR
Specifications
For Use With (Equipment) | CPA Balance, Extended Balance, SE and ME Ultramicro Balance |
---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10465765
![]() |
Sartorius
YSTP01 |
Anti-static Pen |
1890.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Anti-static pen—direct the pen at item to be neutralized
- Ionizing blower—point blower toward sample prior to weighing
Specifications
CPA Balance, Extended Balance, SE and ME Ultramicro Balance |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Anti-Static Devices for Balances