missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ Alkaline Phosphatase
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand: Invitrogen™ IVGN2208
Includes: Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• Utilizes the Anza 10X Buffer
• One-step digestion and dephosphorylation protocol
• Two-step dephosphorylated protocols available
• Heat inactivated in 5 minutes
Certifications
CoA

Specifications
2 mL Anza Alkaline Phosphatase 1 mL Anza Buffer (10X) 1 mL Anza Red Buffer (10X) Store at -5 to -30°C. |
|
Alkaline Phosphatase | |
2000 Reactions | |
Anza™ |
Approved for shipment on Wet or Dry Ice | |
10X Buffer | |
Alkaline Phosphatase |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Insert: Anza Alkaline Phosphatase
Version: MAN0014313 Rev B (6 May 2016)
File size: 158.6 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2814634 | Certificate of Analysis | 29/01/2025 | IVGN2208 |
3069863 | Certificate of Analysis | 27/01/2025 | IVGN2208 |
3049567 | Certificate of Analysis | 25/01/2025 | IVGN2208 |
3064864 | Certificate of Analysis | 25/01/2025 | IVGN2208 |
2781145 | Certificate of Analysis | 09/08/2023 | IVGN2208 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Anza™ Alkaline Phosphatase > No. of Reactions: 2000
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction