missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha/beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA599259
320.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Antibody detects endogenous levels of total AP2 alpha/beta.
The protein encoded by this gene is a transcription factor that binds the consensus sequence 5'-GCCNNNGGC-3'. The encoded protein functions as either a homodimer or as a heterodimer with similar family members. This protein activates the transcription of some genes while inhibiting the transcription of others. Defects in this gene are a cause of branchiooculofacial syndrome. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
AP2 alpha/beta | |
Polyclonal | |
Unconjugated | |
TFAP2B | |
activating enhancer binding protein 2 beta, activating enhancer-binding protein 2 alpha, activating enhancer-binding protein 2 beta, activating enhancer-binding protein 2-alpha, Activating enhancer-binding protein 2-beta, activator protein 2, AI606113, Ap2, AP-2, Ap-2 (a), AP-2 transcription factor, AP-2(beta), AP2alpha, AP-2alpha, AP2-alpha, AP-2B, AP2-B, AP2-beta, AP2TF, BOFS, E130018K07Rik, FLJ51761, Tcfap2a, Tcfap2b, TFAP2, Tfap2a, TFAP2B, transcription factor AP-2 alpha, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2 beta, transcription factor AP-2 beta (activating enhancer binding protein 2 beta), transcription factor AP-2, alpha, transcription factor AP-2-alpha, Transcription factor AP-2-beta | |
Rabbit | |
Affinity chromatography | |
RUO | |
21418, 21419, 301285, 306862, 7020, 7021 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05549, P34056, P58197, Q61313, Q92481 | |
TFAP2A, TFAP2B | |
A synthesized peptide derived from human TFAP2A(Accession P05549), corresponding to amino acid residues K411-K437. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AP2 alpha/beta Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction