Learn More
Applied Biosystems™ MeltDoctor™ HRM Calibration Plate, Fast 96-well
These plates provide thermal and optical calibration required for accurate and reproducible discrimination between melt curves.
Brand: Applied Biosystems™ 4425618
553.36 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: Contains: 1 plate, 96-well fast format
Description
Applied Biosystem™ ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation.
Ready-to-use MeltDoctor HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation.
Fast Real-Time PCR, Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, High Resolution Melting (HRM) Analysis, Methylation Analysis, Methylation-Sensitive HRM Analysis, PCR & Real-Time PCR, RNAi, Epigenetics & Noncoding RNA Research, Real Time PCR (qPCR)
Order Info
Dry ice shipping
Specifications
96-well Plate | |
HRM (High Resolution Melting) Dye | |
Fast | |
HRM Calibration Plate | |
Dry Ice | |
Calibration Plate |
1 plate, 96-well fast format Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
7500 Fast System, 7900HT Fast System | |
MeltDoctor™ | |
Real Time PCR (qPCR) | |
1 x 96-well plate | |
Thermal Calibration, Optic Calibration |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2401102 | Certificate of Analysis | 28/01/2025 | 4425618 |
2408103 | Certificate of Analysis | 26/01/2025 | 4425618 |
2410105 | Certificate of Analysis | 25/01/2025 | 4425618 |
2409104 | Certificate of Analysis | 14/10/2024 | 4425618 |
2312101 | Certificate of Analysis | 25/01/2024 | 4425618 |
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now