Learn More
BRAND™ AR-GLAS™ Stirring Rods
Constructed of AR-GLAS™. Tubes are fused closely at both ends.
Brand:  BRAND™ 135820
55.98 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10597641
  • 71.00 EUR / Pack of 50
Estimated Shipment: 15-04-2025
Description
  • Constructed of AR-GLAS™
  • Tubes are fused closely at both ends.
  • Supplied in packs of 50
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 mm
250 mm
Glass
50/Pk.
Product Title
BRAND™ AR-GLAS™ Stirring Rods > Length: 250mm