Learn More
Tocris Bioscience™ Arformoterol Tartrate
Low molecular weight, organic compound that may help regulate biological process.
Brand:  Tocris Bioscience™ 6219/10
 View more versions of this product
Product Code. 18762449
  • 148.00 EUR / 10mg
Explore more special offers
Description
Description
Arformoterol tartrate is a long-acting β2 adrenergic agonist (LABA). Bronchodilator.
Specifications
Specifications
Long-acting Agonist (LABA)
200815-49-2
494.49g/mol
Adrenergic beta-2 Receptor Agonists
Adrenergic beta-2
C19H24N2O4.C4H6O6
Store at 4°C