missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Arformoterol Tartrate

Low molecular weight, organic compound that may help regulate biological process.

148.00 EUR - 549.00 EUR

Specifications

Product Type Long-acting Agonist (LABA)
Target Proteins Adrenergic beta-2
CAS 200815-49-2
Molecular Formula C19H24N2O4.C4H6O6
Formula Weight 494.49g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18762449
View Documents
Tocris Bioscience™
6219/10
10 mg
148.00 EUR
10mg
Please to purchase this item. Need a web account? Register with us today!
18789797
View Documents
Tocris Bioscience™
6219/50
50 mg
549.00 EUR
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Long-acting β2 adrenergic agonist (LABA). Bronchodilator.
Specifications

Specifications

Long-acting Agonist (LABA)
200815-49-2
494.49g/mol
Adrenergic beta-2 Receptor Agonists
Adrenergic beta-2
C19H24N2O4.C4H6O6
Store at 4°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.