missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenazo III disodium salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Arsenazo III disodium salt is a diazo dye reagent used in the spectrophotometric determination of aluminum, gallium and indium. Further, it is used in the complexometric determination of lanthanides, thallium, scandium thorium, uranium and yttrium. In addition to this, it acts as a metal indicator and form specifically colored metal ion complexes in aqueous media.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Arsenazo III disodium salt is a diazo dye reagent used in the spectrophotometric determination of aluminum, gallium and indium. Further, it is used in the complexometric determination of lanthanides, thallium, scandium thorium, uranium and yttrium. In addition to this, it acts as a metal indicator and form specifically colored metal ion complexes in aqueous media.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >320°C |
Quantity | 1 g |
UN Number | UN3465 |
Solubility Information | Soluble in water. |
Formula Weight | 820.34 |
Chemical Name or Material | Arsenazo III disodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'