Learn More
Atipamezole hydrochloride, MedChemExpress
MedChemExpress Atipamezole (MPV-1248) hydrochloride is a potent α2-adrenoceptor antagonist with a Ki of 1.6 nM.

133.65 EUR - 434.70 EUR
Specifications
Chemical Name or Material | Atipamezole hydrochloride |
---|---|
CAS | 104075-48-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C14H17ClN2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16475128
![]() |
Medchem Express
HY-12380 |
10 mg |
133.65 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421367
![]() |
Medchem Express
HY-12380 |
50 mg |
434.70 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Atipamezole (MPV-1248) hydrochloride is a potent α2-adrenoceptor antagonist with a Ki of 1.6 nM.Specifications
Atipamezole hydrochloride | |
White | |
C14H17ClN2 | |
H2O : 75 mg/mL (301.51 mM; Need ultrasonic and warming) ∣DMSO : ≥ 47 mg/mL (188.94 mM) | |
248.75 | |
97.12% | |
Solid |
104075-48-1 | |
Research | |
MPV-1248 hydrochloride | |
CCC1(C2=CN=CN2)CC3=C(C=CC=C3)C1.[H]Cl | |
248.75 | |
Research | |
Neuroscience-Neuromodulation |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.