Learn More
Thermo Scientific™ Atrial Natriuretic Peptide (1-29), Chicken
For cellular and molecular biology applications
322.00 EUR - 432.00 EUR
Specifications
Species | Chicken |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Lyophilized Powder |
Molecular Weight (g/mol) | 3160.61 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chicken | |
-30°C to -10°C | |
3160.61 | |
RUO - research use only |
HPLC | |
Lyophilized Powder | |
Atrial Natriuretic Peptide (1-29), chicken |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.