Learn More
3M™ Aura™ Particulate Respirator, FFP3, Unvalved, 9330+
3M™ Aura™ Particulate Respirators 9300+ Series has an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media.
Brand: 3M™ 7000034741
67.66 EUR valid until 2025-06-30
Use promo code "24304" to get your promotional price.
This item is not returnable.
View return policy
Description
- Use 3M™ Aura™ Particulate Respirators 9300+ Series for comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
- The 9300+ series offers FFP3 protection and allows you to talk and work in comfort.
- The 3-panel design allows for greater facial movement, while the sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
- The low resistance filter technology helps you breathe easily and the innovative chin tab allows you to make simple adjustments.
- Some of the range features a 3M™ Cool Flow™ Valve to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
- They're individually packed in hygienic wrapping to prevent contamination before use, and the braided, color-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
- Their foldable design makes them easy to store when not in use, and they work well with 3M Eyewear and Hearing Protection products.
Specifications
White | |
Single-use | |
Composite Fabric | |
One Size |
Flat-fold | |
FFP3 | |
General Purpose Particulate Respirator | |
No |
Product Content Correction
Product Title
3M™ Aura™ Particulate Respirator, FFP3, Unvalved, 9330+ > Color: White
Spot an opportunity for improvement?Share a Content Correction