Learn More
Axygen™ Maximum Recovery™ Polypropylene Pipette Tip
For ultra-low retention and high yield
56.31 EUR - 338.53 EUR
Specifications
Sterility | Sterile |
---|---|
Clean Claims | DNase, RNase, and Pyrogen Free |
Format | Rack |
For Use With (Equipment) | Universal Fit |
Product Line | Maxymum Recovery |
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||||
12649535
|
Axygen™
T-200-C-L-R-S |
1 to 200 μL | Clear |
88.23 EUR
Pack of 960 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||||
12679595
|
Axygen™
T-200-Y-R |
200 μL | Yellow |
56.31 EUR
Pack of 960 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||||
12786599
|
Axygen™
T-300-L-R-S |
0.5 to 10 μL | Clear | N/A | N/A | N/A | |||||
11376054
|
Axygen™
T-300-L-R-S |
0.5 to 10 μL | Clear |
92.18 EUR
Pack of 960 |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||||
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Beveled
- Maxymum Recovery technology for ultra-low retention and high yield
- Sterile
Specifications
Sterile | |
Rack | |
Maxymum Recovery |
DNase, RNase, and Pyrogen Free | |
Universal Fit | |
Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.