Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Brand: Axygen™ HTF-1000-CBK-HTR-S
200.30 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.
The tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- RNase-/DNase-free and non-pyrogenic
- Available in non-sterile, sterile, non-filtered and filtered
- Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
Specifications
1000 μL | |
Sterile | |
Not Autoclavable | |
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
Automation, Round | |
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus |
Filtered | |
Black | |
DNase, RNase and Pyrogen free | |
Hanging Tip Rack | |
1 Rack/Pk, 16 Pks/Cs. | |
Polypropylene |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
15Minutes
50Seconds
27