missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates

For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-DW-10ML-24-C-S
This item is not returnable.
View return policy
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
Standard | |
Autoclavable | |
Axygen | |
Microplate | |
10,000 μL | |
Sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
25/Cs. | |
-112°F |
4 x 6 | |
25 Cases | |
Clear | |
Polypropylene | |
24 | |
Square | |
4000 x g | |
5/Pk. | |
-80°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
33024067 | Certificate of Quality | 26/03/2025 | P-DW-10ML-24-C-S |
32824041 | Certificate of Quality | 26/03/2025 | P-DW-10ML-24-C-S |
32924017 | Certificate of Quality | 20/03/2025 | P-DW-10ML-24-C-S |
32724055 | Certificate of Quality | 04/03/2025 | P-DW-10ML-24-C-S |
32424129 | Certificate of Quality | 04/03/2025 | P-DW-10ML-24-C-S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ Storage Microplates > 24-well; 10 mL; Yes; Square
Spot an opportunity for improvement?Share a Content Correction