Learn More
Tocris Bioscience™ AZD 7762 hydrochloride
Potent and selective ATP-competitive inhibitor of Chk1 and Chk2
Brand: Tocris Bioscience™ 5199/50
This item is not returnable.
View return policy
Description
AZD 7762 hydrochloride is a potent and selective ATP-competitive inhibitor of Chk1 and Chk2 (IC50 vales are 5 nM for both kinases); displays at least >10 fold selectivity over a panel of 164 kinases. Potentiates cytotoxicity of DNA-damaging agents. Active in vivo. Also improves efficiency of CRISPR-Cpf1-mediated genome editing in hPSC lines (2.7-fold at 1 μM).Specifications
>98% | |
AZD 7762 hydrochloride | |
AZD 7762 hydrochloride |
50 mg | |
398.88 |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction