Learn More
Tocris Bioscience™ AZD 7762 hydrochloride
Potent and selective ATP-competitive inhibitor of Chk1 and Chk2
Brand:  Tocris Bioscience™ 5199/10
 View more versions of this product
Product Code. 18791548
  • 204.00 EUR / 10mg
[resources not loaded]
Explore more special offers
Description
Description
AZD 7762 hydrochloride is a potent and selective ATP-competitive inhibitor of Chk1 and Chk2 (IC50 vales are 5 nM for both kinases); displays at least >10 fold selectivity over a panel of 164 kinases. Potentiates cytotoxicity of DNA-damaging agents. Active in vivo. Also improves efficiency of CRISPR-Cpf1-mediated genome editing in hPSC lines (2.7-fold at 1 μM).
Specifications
Specifications
>98%
AZD 7762 hydrochloride
AZD 7762 hydrochloride
10 mg
398.88