Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels with Vial Top
Nylon cloth labels for BBP33/i3300 printers.
Brand: Brady™ 143011
This item is not returnable.
View return policy
Description
For BBP33/i3300 printers.
Specifications
UL Recognized,CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only | |
Nylon Cloth Labels for BBP33/i3300 Printers | |
White | |
Nylon Cloth | |
Non-sterile | |
BBP33, i3300,i5300 | |
1500 Stickers | |
-40°F to +194°F | |
0.0065 in. | |
22.86 mm (W) x 12.70 mm (H) x 12.70 mm (Dia) | |
12.70 mm | |
22.86 mm | |
12.70 mm |
B33-126-499 | |
Permanent Acrylic | |
Matte | |
Labels | |
Laboratory labelling | |
1 | |
Rectangle | |
-40°C to +90°C | |
0.166 mm | |
0.5 in. | |
0.9 in. | |
0.5 in. |
Product Content Correction
Product Title
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels with Vial Top > Brady™ Nylon Cloth Labels
Spot an opportunity for improvement?Share a Content Correction