Learn More
Saint Gobain Life Sciences BarbLock™ Hand Tool
BarbLock™ Hand Tool with stainless steel jaw and handle installs BarbLock™ retainers to tubing.
Brand:  Saint Gobain Life Sciences BLT135125
 View more versions of this product
Product Code. 30040264
  • 310.00 EUR / Each
Explore more special offers
Description
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Clamp
125 Series
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Saint Gobain Life Sciences BarbLock™ Hand Tool > 125 Series