Learn More
Eppendorf™ Battery for Multipette Stream and Xstream
Use this battery to charge Eppendorf™ Multipette E3/E3x and Multipette stream/Xstream electronic dispensers.
Brand: Eppendorf™ 4986602009
18.98 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Replacement battery for Eppendorf Multipette E3/E3x and Multipette stream/Xstream
- 3.7 Volts
Specifications
Li-ion | |
1 Each | |
Battery | |
Multipette Stream and Xstream |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
4Hours
13Minutes
15Seconds
52 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
21Hours
13Minutes
15Seconds
52