Learn More
BD™ Bactec™ Media: Lytic/10 Anaerobic/F
For use with BACTEC 9240, 9120, and 9050 Systems
Brand:  BD™ 442265
Product Code. 15968041
Description
Description
  • Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
  • Fluorescent series bar-coded blood culture media
  • Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
  • Vials ready to use as received—require no reconstitution or dilution
  • Detection is limited to microorganisms that will grow in a particular type of medium
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 25°C
Lytic/10 anaerobic/F
Processed Water 40mL, Soybean Casein digest broth 2.75% w/v, Yeast extract 0.2% w/v, Animal Tissue digest 0.05% w/v, Dextrose 0.2% w/v, Hemin 0.0005% w/v, Menadione 0.00005% w/v, Thiols 0.10% w/v, Sodium Pyruvate 0.1% w/v, Saponin 0.26% w/v, Antifoaming Agent 0.01% w/v, Sodium Polyanetholsulfonate (SPS) 0.035% w/v
Vial
Liquid
Microbiology Media
Bactec™ 9240, 9120 and 9050 System
50/Pk.
Documents
One moment while we fetch your results.
Product Title
BD™ Bactec™ Media: Lytic/10 Anaerobic/F > BACTEC Lytic/10 Anaerobic/F Medium; 50/Pk.