Learn More
BD BBL™ Mycoflask™ Prepared Pour Tubed Media: Trypticase™ Soy Agar (Blood Agar Base) Deeps
For the isolation and cultivation of fastidious and nonfastidious anaerobic and aerobic microorganisms
Brand:  BD 221082
Product Code. 17302321
  • 55.25 EUR / Pack of 10
Explore more special offers
Description
Description
  • For cultures requiring prolonged incubation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Liquid
Trypticase soy agar
Isolation and cultivation of non-fastidious and fastidious microorganisms
20 mL
Beige
United States
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD BBL™ Mycoflask™ Prepared Pour Tubed Media: Trypticase™ Soy Agar (Blood Agar Base) Deeps > Trypticase soy agar (blood agar base) deeps; 20mL