Learn More
BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin
Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials
Brand: BD 221788
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, BS12, CLSI, CMPH2, MCM9 | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L | |
Size K tube with tight cap |
Light Yellow | |
Liquid | |
Thioglycollate Medium | |
Isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials | |
100 x 8 mL |
Product Suggestions
Customers who viewed this item also viewed.
For in vitro diagnostic use.