Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefoxitin20
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
189.00 EUR
Specifications
Antimicrobial Agents Cefoxitin20
Concentration 30μg
Packaging Type Disc
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17362381
View Documents
BD
231591
10 Pack
189.00 EUR
Pack of 10
Estimated Shipment: 02-01-2025
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Cefoxitin20
30μg
Disc
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 913 Days
SDS
Documents
Documents
Product Certifications
For in vitro diagnostic use