Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefoxitin20
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
189.00 EUR
Specifications
Antimicrobial Agents | Cefoxitin20 |
---|---|
Concentration | 30μg |
Packaging Type | Disc |
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Cefoxitin20 | |
30μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 913 Days
For in vitro diagnostic use