Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefoxitin20
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand:  BD 231591
 View more versions of this product
Product Code. 17362381
  • 189.00 EUR / Pack of 10
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cefoxitin20
30μg
Disc
10 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefoxitin20 > Quantity: 10/Pk.
For in vitro diagnostic use