Learn More
BD CBA Mouse Inflammation Kit
Optimized for analysis of specific proteins in tissue culture supernatants, EDTA plasma, and serum samples
Brand: BD 552364
1759.02 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-γ, TNF, and IL-12p70; wash buffer; assay diluent; cytometer setup reagents
Description
- Can be used to quantitatively measure Interleukin-6 (IL-6), Interleukin-10 (IL-10), Monocyte Chemoattractant Protein-1 (MCP-1), Interferon-γ (IFN-γ), Tumor Necrosis Factor (TNF), and Interleukin-12p70 (IL-12p70) protein levels in a single sample
- Detection reagent provided in the kit is a mixture of phycoerythrin (PE)-conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
- Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
IFN-gamma | |
Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-g, TNF and IL-12p70, Wash Buffer, assay diluent, cytometer setup reagents | |
2°C to 8°C | |
Mouse inflammation kit |
Mouse | |
80 Tests | |
Flow Cytometry |
Product Content Correction
Product Title
BD CBA Mouse Inflammation Kit > 1 Kit (80 tests)
Spot an opportunity for improvement?Share a Content Correction