Learn More
BD Cytometric Bead Array (CBA) Human Anaphylatoxin Kit
Can be used to quantitatively measure anaphylatoxin C3a, C4a, and C5a (bioactive cleavage products released from C3, C4, and C5 during complement activation) protein levels in a single EDTA plasma or serum sample.
Brand: BD 561418
1561.07 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Performance has been optimized for analysis of specific proteins in EDTA plasma and serum samples
- Provides sufficient reagents for the quantitative analysis of 80 samples
Applications:
Flow Cytometry
Specifications
Anaphylatoxin | |
Reagents for capture and detection of C4a, C3a and C5a, standards, Wash Buffer, assay diluent, cytometer setup reagents | |
2°C to 8°C | |
Human anaphylatoxin kit |
Human | |
25 Tests | |
Flow Cytometry |