missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD DAB Substrate Kit
For immunoassay protocols
Brand: BD 550880
Includes: DAB buffer (50mL) is a solution of citric acid and H2O2. DAB chromagen (2.5mL) is a solution of 3-3 Diaminobenzidine(1.74% w/v) and HCl.
Description
- Substrate for enzyme peroxidase
- Chromogen
- Produces brown precipitate insoluble in alcohol
Immunohistochemistry-frozen/paraffin

Specifications
2.5mL | |
Immunohistochemistry frozen or paraffin | |
50mL DAB buffer is a solution of citric acid and H2O2, 2.5mL DAB chromagen, is a solution of 1.74% w/v 3-3 Diaminobenzidine and HCl | |
4°C |
DAB | |
DAB Substrate | |
7.2 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD DAB Substrate Kit > 1 kit
Research use only
Spot an opportunity for improvement?Share a Content Correction