missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD OptEIA™ Human IL-15 ELISA Set
For developing ELISA for natural or recombinant human IL-15 in serum, plasma and cell culture supernatants
Brand: BD 559268
989.11 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Capture antibody, detection antibody, streptavidin-HRP standards.
Description
- Size: 20 Plates

Specifications
ELISA Kit | |
Human | |
ELISA | |
2°C to 8°C |
20 Plates | |
Liquid | |
Capture Antibody, detection Antibody, Streptavidin-HRP, standards |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD OptEIA™ Human IL-15 ELISA Set > Human IL-15 ELISA Set; 20 Plates
Spot an opportunity for improvement?Share a Content Correction