missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Rat Cerebrum Lysate
Convenient to use and useful control for western blotting application
Brand: BD 611463
This item is not returnable.
View return policy
Description
- Source is rich in endogenously expressed proteins
- Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
- Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
Routinely tested western blot

Specifications
Cerebrum | |
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol | |
1 mg/mL |
500 μg | |
Rat | |
-20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Rat Cerebrum Lysate > 500μg
Research use only
Spot an opportunity for improvement?Share a Content Correction