missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
CAS | 119-53-9 |
Molecular Formula | C14H12O2 |
MDL Number | MFCD00004496 |
Quantity | 100 g |
Synonym | benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone |
InChI Key | ISAOCJYIOMOJEB-UHFFFAOYNA-N |
SMILES | OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
IUPAC Name | 2-hydroxy-1,2-diphenylethan-1-one |
Molecular Weight (g/mol) | 212.25 |
PubChem CID | 8400 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Benzoin, MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction