missing translation for 'onlineSavingsMsg'
Learn More
Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals Product Code.: 15468317

Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 15468317
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15468317

Brand: Thermo Scientific Alfa Aesar H51844.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32894-07-8
Molecular Formula C9H12ClNS
Molecular Weight (g/mol) 201.712
MDL Number MFCD05865210
InChI Key OVQWHRFBVPDPCD-UHFFFAOYSA-N
Synonym s-benzylthioacetimidate, hydrochloride, benzyl thioacetimidate hydrochloride, benzyl thioacetimidate hcl, benzyl ethanimidothioate hydrochloride, 1-benzylsulfanyl ethanimine hydrochloride, acmc-20ajr8, s-benzylthioacetimidate hydrochloride, thioacetimidic acid benzyl ester; hydrochloride, benzyl ethanimidothioate-hydrogen chloride 1/1Show MoreShow Less
PubChem CID 12626231
IUPAC Name benzyl ethanimidothioate;hydrochloride
SMILES CC(=N)SCC1=CC=CC=C1.Cl

Specifications

Quantity 5 g
Formula Weight 201.71
Percent Purity 96%
Chemical Name or Material Benzyl thioacetimidate hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10168289 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberH51844.06,H51844.03,H51844.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.