Learn More
Invitrogen™ Human Betacellulin ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand:  Invitrogen™ 900-K372
Product Code. 17889025
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P35070
HRP
Colorimetric Microplate Reader
BTC
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
1 hrs 45 mins
Lot-specific
ELISA
685
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
RUO
Betacellulin
24 hrs 30 mins
Product Title
Invitrogen™ Human Betacellulin ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP