Learn More
Invitrogen™ Human Betacellulin ELISA Development Kit (ABTS), PeproTech®
ELISA
Brand: Invitrogen™ 900-K372
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
Specifications
P35070 | |
HRP | |
Colorimetric Microplate Reader | |
BTC | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 45 mins |
Lot-specific | |
ELISA | |
685 | |
Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate | |
RUO | |
Betacellulin | |
24 hrs 30 mins |
Product Content Correction
Product Title
Invitrogen™ Human Betacellulin ELISA Development Kit (ABTS), PeproTech® > 10 x 96 Tests; HRP
Spot an opportunity for improvement?Share a Content Correction