missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Bile bovine, powder Product Code.: 15517529

Thermo Scientific Chemicals Bile bovine, powder

Product Code. 15517529
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15517529

Brand: Thermo Scientific Chemicals 460331000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-63-7
Molecular Formula C24H40O5
Molecular Weight (g/mol) 408.58
InChI Key BHQCQFFYRZLCQQ-UHFFFAOYNA-N
IUPAC Name 4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoic acid
SMILES CC(CCC(O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C

Specifications

Quantity 100 g
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Bile bovine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463037 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number460331000,460330250,460335000
Lot NumberA0451581 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number460331000,460330250,460335000
Lot NumberA0436166 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number
Lot NumberA0408800 Certificate TypeCertificate of Analysis Date02/12/2020 Catalog Number460331000,460330250,460335000
Lot NumberA0424337 Certificate TypeCertificate of Analysis Date19/11/2020 Catalog Number460331000,460330250,460335000
Product Identifier
  • Bile bovine, powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Bile bovine, powder >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.